Learn More
Aviva Systems Biology Corp AKT1 Antibody - N-terminal region (AVARP06008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP06008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AKT1. Alternate names: AKT, PKB, RAC, PRKBA, PKB-ALPHA, RAC-ALPHAApplicationWB. Species reactivity: Cow, Dog, Goat, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-62540