Learn More
Sino Biological Recombinant Human ACVR1 Protein (R258G, GST Tag), Active
Catalog No. 502576042
Encompass_Preferred
Recombinant human ALK2 (R258G) (147-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-6042 Supplier Sino Biological Supplier No. A0612KG500
Recombinant human ALK2 (R258G) (147-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.