Learn More
Enzo Life Sciences Akt1 (human), (recombinant) (GST-tag) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4160005
Encompass_Preferred
Alternative name: Protein kinase B alpha, PKB alpha, RAC alpha serine/threonine protein kinase. Source: Produced in insect cells. Full length active enzyme. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2017