Learn More
Medchemexpress LLC Akt1 protein, human (active, Sf9, GST) | 100UG
SDP
Catalog No. 5000176634
Encompass_Preferred
Recombinant human AKT1 protein (active) expressed in Sf9 insect cells with an N-terminal GST tag. Supplied as a 0.22 μm filtered solution formulated in 50 mM HEPES (pH 7.5), 200 mM NaCl, 20% glycerol, 5 mM DTT, and trehalose; intended for research applications requiring active AKT1 kinase.
  • Active kinase suitable for activity assays and signaling studies.
  • Full-length sequence (M1-A480), approximately 82.2 kDa.
  • N-terminal GST tag facilitates purification and detection.
  • Expressed in Sf9 insect cells to support eukaryotic modifications.
  • Supplied as a filtered solution in stabilizing buffer for handling and storage.
  • Recommend aliquoting and avoiding repeated freeze-thaw cycles to preserve activity.
Catalog No. 50-001-76634 Supplier Medchemexpress LLC Supplier No. HYP702631100UG
Recombinant human AKT1 protein (active) expressed in Sf9 insect cells with an N-terminal GST tag. Supplied as a 0.22 μm filtered solution formulated in 50 mM HEPES (pH 7.5), 200 mM NaCl, 20% glycerol, 5 mM DTT, and trehalose; intended for research applications requiring active AKT1 kinase.
  • Active kinase suitable for activity assays and signaling studies.
  • Full-length sequence (M1-A480), approximately 82.2 kDa.
  • N-terminal GST tag facilitates purification and detection.
  • Expressed in Sf9 insect cells to support eukaryotic modifications.
  • Supplied as a filtered solution in stabilizing buffer for handling and storage.
  • Recommend aliquoting and avoiding repeated freeze-thaw cycles to preserve activity.