Learn More
Aviva Systems Biology Corp AKT1S1 Antibody - C-terminal region (ARP48061_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48061_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKT1S1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human AKT1S1. Alternate names: Lobe, MGC2865, PRAS40ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53537