Learn More
Carnabio Usa Inc AKT2/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0110210
Encompass
AKT2, Catalytic domain, Wild type, Amino Acid 120-481, NP_001617.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0675