Learn More
Aviva Systems Biology Corp AKT2 Antibody - middle region (ARP30321_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30321_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AKT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AKT2. Alternate names: PKBBETA, PRKBB, RAC-BETA, PKBB, HIHGHHApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-54155