Learn More
Medchemexpress LLC AKT2 Human sf9 Hi 5ug
SDP
Catalog No. 5000178899
Encompass_Preferred
The AKT2 protein is a serine/threonine protein kinase in the AKT family that is critical for metabolism proliferation survival growth and angiogenesis It phosphorylates more than 100 substrates coordinates insulin-induced SLC2A4/GLUT4 translocation affects glucose uptake and controls glycogen storage AKT2 Protein Human (sf9 His-GST) is the recombinant human-derived AKT2 protein expressed by Sf9 insect cells with C-His N-GST labeled tag
Catalog No. 50-001-78899 Supplier Medchemexpress LLC Supplier No. HYP744205UG
The AKT2 protein is a serine/threonine protein kinase in the AKT family that is critical for metabolism proliferation survival growth and angiogenesis It phosphorylates more than 100 substrates coordinates insulin-induced SLC2A4/GLUT4 translocation affects glucose uptake and controls glycogen storage AKT2 Protein Human (sf9 His-GST) is the recombinant human-derived AKT2 protein expressed by Sf9 insect cells with C-His N-GST labeled tag