Learn More
Sino Biological Recombinant Human AKT1 Protein (Full Length, E17K, GST Tag)
Catalog No. 502575995
Encompass_Preferred
Recombinant full length human AKT1 (E17K) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5995 Supplier Sino Biological Supplier No. A1616G100
Recombinant full length human AKT1 (E17K) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.