Learn More
ABclonal Technology AKT3 Rabbit mAb
SDP
Catalog No. 5000005386
Encompass_Preferred
The protein encoded by this gene is a member of the AKT also called PKB serine threonine protein kinase family AKT kinases are known to be regulators of cell signaling in response to insulin and growth factors They are involved in a wide variety of biological processes including cell proliferation differentiation apoptosis tumorigenesis as well as glycogen synthesis and glucose uptake This kinase has been shown to be stimulated by platelet-derived growth factor (PDGF) insulin and insulin-like growth factor 1 (IGF1) Alternatively splice transcript variants encoding distinct isoforms have been described
Catalog No. 50-000-05386 Supplier ABclonal Technology Supplier No. A22738100UL
The protein encoded by this gene is a member of the AKT also called PKB serine threonine protein kinase family AKT kinases are known to be regulators of cell signaling in response to insulin and growth factors They are involved in a wide variety of biological processes including cell proliferation differentiation apoptosis tumorigenesis as well as glycogen synthesis and glucose uptake This kinase has been shown to be stimulated by platelet-derived growth factor (PDGF) insulin and insulin-like growth factor 1 (IGF1) Alternatively splice transcript variants encoding distinct isoforms have been described