Learn More
Selleck Chemical LLC Akti-1/2 5mg 612847-09-3 Akt Inhibitor VIII
SDP
Supplier:  Selleck Chemical LLC S77765MG
Encompass_Preferred
Akti-1/2 (Akt Inhibitor VIII) is a highly selective Akt1/Akt2 inhibitor with IC50 of 58 nM/210 nM, respectively, about 36-fold selectivity for Akt1 over Akt3. Akti-1/2 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5236