Learn More
Insphero Inc Akura™ 384 Spheroid Microplate - powered by InSphero
SDP
Catalog No. NC2028090
Encompass
The ultimate spheroid screening plate unifying robust liquid and spheroid handling w/ automated high-resolution imaging. The flat bottom and black walls of the InSphero 384 well plate facilitates high-content imaging while the lower cavity of the wells allows medium exchange, dosing & endpoint determination in the same plate saving time and resources.1. Unique SureXchange™ ledge engineered to prevent unintended spheroid/organoid aspiration2. Spheroid/organoid morphology and functionality is preserved in long-term culture3. Superior high content imaging quality with a continuous, flat-bottom, 25 μm, gas-permeable membrane, & a black-walled body4. Improved spheroid/organoid formation with the Akura™ Tilting Stand by keeping plate at 30 degrees during cell aggregation 10-pack of individually wrapped Akura™ 384 Spheroid Microplates w/ black-walled polystyrene body bonded to transparent, continuous, 25 µm
Catalog No. NC2028090 Supplier Insphero Inc Supplier No. CSPC14
The ultimate spheroid screening plate unifying robust liquid and spheroid handling w/ automated high-resolution imaging. The flat bottom and black walls of the InSphero 384 well plate facilitates high-content imaging while the lower cavity of the wells allows medium exchange, dosing & endpoint determination in the same plate saving time and resources.1. Unique SureXchange™ ledge engineered to prevent unintended spheroid/organoid aspiration2. Spheroid/organoid morphology and functionality is preserved in long-term culture3. Superior high content imaging quality with a continuous, flat-bottom, 25 μm, gas-permeable membrane, & a black-walled body4. Improved spheroid/organoid formation with the Akura™ Tilting Stand by keeping plate at 30 degrees during cell aggregation 10-pack of individually wrapped Akura™ 384 Spheroid Microplates w/ black-walled polystyrene body bonded to transparent, continuous, 25 µm