Learn More
Insphero Inc Akura™ 96 Spheroid Microplate - powered by InSphero
SDP
Catalog No. NC2121069
Encompass
Automation compatible – Secure handling – Easy observationThe one-and-only 96-well spheroid plate with InSphero’s unique features specially designed for lossless spheroid/organoid formation and handling as well as reliable culturing and analysis.1. Unique SureXchange™ ledge engineered to prevent unintended spheroid/organoid aspiration2. Spheroid/organoid morphology and functionality is preserved in long-term culture by unique ULA coating3. Enhanced imaging speed and resolution through flat bottom well4. Improved spheroid/organoid formation with the Akura™ Tilting Stand by keeping plate at 30 degrees during cell aggregation5/Pack of individually wrapped Akura™ 96 Spheroid Microplates, made of Cyclo Olefin Polymer, high-efficiency ULA coating, sterile, includes lid.For further information, please visit www.insphero.com
This item is not returnable. View return policy
Catalog No. NC2121069 Supplier Insphero Inc Supplier No. CSPB13
Automation compatible – Secure handling – Easy observationThe one-and-only 96-well spheroid plate with InSphero’s unique features specially designed for lossless spheroid/organoid formation and handling as well as reliable culturing and analysis.1. Unique SureXchange™ ledge engineered to prevent unintended spheroid/organoid aspiration2. Spheroid/organoid morphology and functionality is preserved in long-term culture by unique ULA coating3. Enhanced imaging speed and resolution through flat bottom well4. Improved spheroid/organoid formation with the Akura™ Tilting Stand by keeping plate at 30 degrees during cell aggregation5/Pack of individually wrapped Akura™ 96 Spheroid Microplates, made of Cyclo Olefin Polymer, high-efficiency ULA coating, sterile, includes lid.For further information, please visit www.insphero.com