Learn More
Insphero Inc Akura™ Tilting Stand - powered by InSphero
SDP
Catalog No. NC2044831
Encompass
The Akura™ Tilting Stand conveniently keeps well plates at an angle of 30 degrees during cell aggregation improving spheroid/organoid uniformity.1. Improved spheroid uniformity through tilting of Akura™ 96/384 Spheroid Plates and directing cells to the edge of the well for efficient aggregation2. Safe stacking of up to 4 plates for parallel spheroid/organoid formation3. Incubator safe and reusable through simple cleaning and sterilizationOne stand, individually wrapped, made of anodized aluminum, non-sterile.For further information, please visit www.insphero.com
Catalog No. NC2044831 Supplier Insphero Inc Supplier No. CSAG11
The Akura™ Tilting Stand conveniently keeps well plates at an angle of 30 degrees during cell aggregation improving spheroid/organoid uniformity.1. Improved spheroid uniformity through tilting of Akura™ 96/384 Spheroid Plates and directing cells to the edge of the well for efficient aggregation2. Safe stacking of up to 4 plates for parallel spheroid/organoid formation3. Incubator safe and reusable through simple cleaning and sterilizationOne stand, individually wrapped, made of anodized aluminum, non-sterile.For further information, please visit www.insphero.com