Learn More
Medchemexpress LLC HY-15709 5mg Medchemexpress, AL 082D06 CAS:256925-03-8 Purity:>98%
SDP
Catalog No. 502081774
Encompass_Preferred
Medchemexpress, HY-15709 5mg AL 082D06 CAS:256925-03-8 AL 082D06 is a selective, nonsteroidal glucocorticoid receptor (GR) antagonist with Ki of 210 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1774 Supplier Medchemexpress LLC Supplier No. HY157095MG
Medchemexpress, HY-15709 5mg AL 082D06 CAS:256925-03-8 AL 082D06 is a selective, nonsteroidal glucocorticoid receptor (GR) antagonist with Ki of 210 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.