Learn More
Biosurface Technologies Corp Dual Channel Transmission Flow Cell (aluminum flow channel; microtiter plate footprint), for Growing and Imaging of Biofilms.
Catalog No. NC3662472
Encompass
"The models FC 280 and 281 flow cell design is based on the FC 81 Single Channel Transmission Flow Cell but accommodates two growth channels. This design allows biofilms to be grown side by side for duplicate or “control–treatment” experiments. Biofilms growing in each channel can be easily compared microscopically by simply moving the microscope stage backward or forwards. The body of the flow cell consists of a polycarbonate flow channel. Glass viewing ports allow transmitted, reflected, fluorescent, and confocal microscopy of biofilm growth in the flow channels. All units are autoclavable and re-useable.The viewing windows consist of a no.2, 24x60 mm cover slip. The opposing viewing window consists of a standard 75 x 25x 1 mm glass microscope slide. The glass viewing windows are held in place by an aluminum cover plate. The cover plates also compress the silicone rubber gasket to provide a leak-proof flow cell.
Catalog No. NC3662472 Supplier Biosurface Technologies Corp Supplier No. FC 281AL
"The models FC 280 and 281 flow cell design is based on the FC 81 Single Channel Transmission Flow Cell but accommodates two growth channels. This design allows biofilms to be grown side by side for duplicate or “control–treatment” experiments. Biofilms growing in each channel can be easily compared microscopically by simply moving the microscope stage backward or forwards. The body of the flow cell consists of a polycarbonate flow channel. Glass viewing ports allow transmitted, reflected, fluorescent, and confocal microscopy of biofilm growth in the flow channels. All units are autoclavable and re-useable.The viewing windows consist of a no.2, 24x60 mm cover slip. The opposing viewing window consists of a standard 75 x 25x 1 mm glass microscope slide. The glass viewing windows are held in place by an aluminum cover plate. The cover plates also compress the silicone rubber gasket to provide a leak-proof flow cell.