Learn More
Biosurface Technologies Corp Dual Channel Coupon Evaluation Flow Cell (aluminum flow channel; platform stage mount), for Growing and Imaging of Biofilms on Disc Coupons.
Catalog No. NC3662438
Encompass
"The models FC 270 and 271 flow cell design is based on the FC 71 but accommodates two growthchannels. This allows biofilms to be grown side by side for duplicate or “control – treatment” experiments. Biofilms growing in each channel can be easily compared microscopically by simply moving the microscope stage backwards or forwards. The body of the flow cell consists of a polycarbonate or anodized aluminum flow channel. Glass viewing ports allow reflected, fluorescent, and confocal microscopy of biofilm growth in the flow channels.The viewing window consists of a no. 2, 24x60 mm cover slip. The glass viewing window is held in place by an aluminum cover plate. The cover plates also compress the silicone rubber gasket to provide a leak-proof flow cell.The flow channel has been recessed to accept standard FC 71 coupon sizes (one 6x25x2 mm thick; three 10 mm diameter x 2 mm thick disc coupons.
Catalog No. NC3662438 Supplier Biosurface Technologies Corp Supplier No. FC 270AL2X0.5
"The models FC 270 and 271 flow cell design is based on the FC 71 but accommodates two growthchannels. This allows biofilms to be grown side by side for duplicate or “control – treatment” experiments. Biofilms growing in each channel can be easily compared microscopically by simply moving the microscope stage backwards or forwards. The body of the flow cell consists of a polycarbonate or anodized aluminum flow channel. Glass viewing ports allow reflected, fluorescent, and confocal microscopy of biofilm growth in the flow channels.The viewing window consists of a no. 2, 24x60 mm cover slip. The glass viewing window is held in place by an aluminum cover plate. The cover plates also compress the silicone rubber gasket to provide a leak-proof flow cell.The flow channel has been recessed to accept standard FC 71 coupon sizes (one 6x25x2 mm thick; three 10 mm diameter x 2 mm thick disc coupons.