Learn More
Al-23 Combustion Tray;L1 (mm), 100;L2 (mm), 98;B1 (mm), 50;B2 (mm), 48;H (mm), 30;W (mm), 4;Vol, 100.0
Catalog No. AA33186KF
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
AA33186KF 1 Ea.
1 options
Catalog No. AA33186KF Supplier Thermo Scientific Chemicals Supplier No. 033186KF
Add to Cart
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Combustion Tray
Quantity 1 Ea.
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.