Learn More
Cayman Chemical AL 34662 (formate), Purity: ≥98%, 10 mg
SDP
Catalog No. 502944635
Encompass_Preferred
A 5-HT2 receptor agonist; selectively binds to 5-HT2 receptors (IC50 = 1.5 and 0.77 nM in isolated and washed human or rat cerebral cortex homogenates, respectively) over a panel of 19 neurotransmitter and steroid receptors at 10 µM; induces calcium mobilization in a reporter assay using primary human ciliary muscle cells (EC50 = 140 nM); reduces IOP in a cynomolgus monkey model of trabeculoplasty-induced ocular hypertension at 100 or 300 µg/eye
Catalog No. 50-294-4635 Supplier Cayman Chemical Supplier No. 4048910 MG
A 5-HT2 receptor agonist; selectively binds to 5-HT2 receptors (IC50 = 1.5 and 0.77 nM in isolated and washed human or rat cerebral cortex homogenates, respectively) over a panel of 19 neurotransmitter and steroid receptors at 10 µM; induces calcium mobilization in a reporter assay using primary human ciliary muscle cells (EC50 = 140 nM); reduces IOP in a cynomolgus monkey model of trabeculoplasty-induced ocular hypertension at 100 or 300 µg/eye