Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AL-611 | 2481279-61-0 | 99.9% | C25H33F2N6O8P | 1 MG
SDP

Catalog No. 5000356003
Encompass_Preferred

AL-611 is an HCV NS5B polymerase inhibitor with an EC50 of 5 nM. It targets HCV Protease and is involved in anti-infection and metabolic enzyme/protease pathways.

  • HCV NS5B polymerase inhibitor (EC50 = 5 nM)
  • Targets HCV Protease
  • Involved in anti-infection and metabolic enzyme/protease pathways
  • Molecular weight: 614.54
  • Soluble in DMSO at 125 mg/mL (requires ultrasonication)
  • Appears as a white to off-white solid
  • Used in research for chronic Hepatitis C treatment

Catalog No. 50-003-56003 Supplier Medchemexpress LLC Supplier No. HY1453741MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AL-611 is an HCV NS5B polymerase inhibitor with an EC50 of 5 nM. It targets HCV Protease and is involved in anti-infection and metabolic enzyme/protease pathways.

  • HCV NS5B polymerase inhibitor (EC50 = 5 nM)
  • Targets HCV Protease
  • Involved in anti-infection and metabolic enzyme/protease pathways
  • Molecular weight: 614.54
  • Soluble in DMSO at 125 mg/mL (requires ultrasonication)
  • Appears as a white to off-white solid
  • Used in research for chronic Hepatitis C treatment

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.