Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AL-611 | 2481279-61-0 | 99.9% | C25H33F2N6O8P | 1 ML
SDP

Catalog No. 5000354196
Encompass_Preferred

AL-611 is an HCV NS5B polymerase inhibitor with an EC50 of 5 nM, intended for research use only.

  • Molecular weight of 614.54.
  • Purity of 99.93%.
  • Appears as a solid, white to off-white.
  • Soluble in DMSO (125 mg/mL).
  • Store powder at -20°C for up to 3 years.
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month.
  • Exhibits CC50 > 100 μM and EC50 0.005 μM in Huh-7 cells.
  • Involved in a Phase 1 clinical trial for Hepatitis C (NCT03253471).

Catalog No. 50-003-54196 Supplier Medchemexpress LLC Supplier No. HY14537410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AL-611 is an HCV NS5B polymerase inhibitor with an EC50 of 5 nM, intended for research use only.

  • Molecular weight of 614.54.
  • Purity of 99.93%.
  • Appears as a solid, white to off-white.
  • Soluble in DMSO (125 mg/mL).
  • Store powder at -20°C for up to 3 years.
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month.
  • Exhibits CC50 > 100 μM and EC50 0.005 μM in Huh-7 cells.
  • Involved in a Phase 1 clinical trial for Hepatitis C (NCT03253471).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.