Learn More
Medchemexpress LLC Al-611 | 2481279-61-0 | 99.9% | C25H33F2N6O8P | 10MG
SDP
Catalog No. 5000260304
Encompass_Preferred
AL-611 is a research-grade small-molecule inhibitor described by the manufacturer as an HCV protease inhibitor intended for laboratory research use. It is supplied as a white to off-white solid and is characterized by a high reported purity and defined molecular properties.
  • Potent HCV protease inhibitor for laboratory research.
  • High purity (99.93%).
  • White to off-white solid, suitable for analytical and biological assays.
  • Molecular weight 614.54 Da; molecular formula C25H33F2N6O8P.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Intended for research use only, not for human or veterinary use.
Catalog No. 50-002-60304 Supplier Medchemexpress LLC Supplier No. HY14537410MG
AL-611 is a research-grade small-molecule inhibitor described by the manufacturer as an HCV protease inhibitor intended for laboratory research use. It is supplied as a white to off-white solid and is characterized by a high reported purity and defined molecular properties.
  • Potent HCV protease inhibitor for laboratory research.
  • High purity (99.93%).
  • White to off-white solid, suitable for analytical and biological assays.
  • Molecular weight 614.54 Da; molecular formula C25H33F2N6O8P.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Intended for research use only, not for human or veterinary use.