Learn More
Medchemexpress LLC HY-108645 5mg Medchemexpress, AL 8697 CAS:1057394-06-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1086455MG
Encompass_Preferred
Medchemexpress, HY-108645 5mg AL 8697 CAS:1057394-06-5 AL 8697, a specific and orally active p38α MAPK inhibitor with an IC 50 of 6 nM, 14-fold less potent in p38β MAPK ( IC 50 =82 nM), exhibits anti-inflammatory activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2328