Learn More
Cayman Chemical AL 8810 ethyl amide
SDP
Catalog No. 502887311
Encompass_Preferred
An N-ethyl amide analog of AL 8810; shows no contraction activity at concentrations up to 0.1 mM and does not antagonize the activity of PGF2α-ethanolamide in cat iris
Catalog No. 50-288-7311 Supplier Cayman Chemical Supplier No. 1001047010 MG
An N-ethyl amide analog of AL 8810; shows no contraction activity at concentrations up to 0.1 mM and does not antagonize the activity of PGF2α-ethanolamide in cat iris