Learn More
Cayman Chemical AL 8810 methyl ester
SDP
Supplier:  Cayman Chemical 100083705 MG
Encompass_Preferred
AL 8810 methyl ester is a lipid soluble, esterified prodrug form of AL 8810 analogous to the commonly used therapeutic intraocular PG compounds such as latanoprost and travoprost. The pharmacology of AL 8810 methyl ester has not been published.
Catalog No. 50-288-7581