Learn More
MSC Simpson Pressure Washer, 4 GPM, 4200 PSI, Gas, Cold Water
Catalog No. 502838848
Encompass
This pressure washer is designed for powerful cleaning applications, featuring a durable pump and a robust hose for extended reach. It is suitable for a variety of tasks requiring high pressure and cold water.
  • Cat triplex pump
  • 50 ft. hose
  • Includes 5 quick-connect nozzles
  • 50 ft. x 3/8 in. steel braided hose
  • Gun and lance
  • Siphon tube
  • Cold water type
  • Gas power type
  • 389 cc displacement
  • 4.0 GPM flow rate
  • 4200 PSI pressure
Catalog No. 50-283-8848 Supplier MSC Supplier No. 49040066
This pressure washer is designed for powerful cleaning applications, featuring a durable pump and a robust hose for extended reach. It is suitable for a variety of tasks requiring high pressure and cold water.
  • Cat triplex pump
  • 50 ft. hose
  • Includes 5 quick-connect nozzles
  • 50 ft. x 3/8 in. steel braided hose
  • Gun and lance
  • Siphon tube
  • Cold water type
  • Gas power type
  • 389 cc displacement
  • 4.0 GPM flow rate
  • 4200 PSI pressure