Learn More
Perkin Elmer US LLC Aluminum (Al) - Calcium (Ca) - Copper (Cu) - Iron (Fe) - Magnesium (Mg) - Silicon (Si) - Zinc (Zn) Lumina Hollow Cathode Lamp
Catalog No. 509044290
Encompass_Preferred
Multi-Element Lumina hollow cathode lamp (HCL) for the detection of elemental Aluminum (Al), Calcium (Ca), Copper (Cu), Iron (Fe), Magnesium (Mg), Silicon (Si), Zinc (Zn). Please note that multi-element lamps are not designed to be used in a furnace system. In addition the analysis of Aluminum and Silicon require the use of a nitrous oxide-acetylene flame. Every genuine PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume — and the longer the lamp lifetime. Note: Lumina hollow cathode lamps may also be used with all earlier AA spectrometers that
Catalog No. 50-904-4290 Supplier Perkin Elmer US LLC Supplier No. N3050218
Multi-Element Lumina hollow cathode lamp (HCL) for the detection of elemental Aluminum (Al), Calcium (Ca), Copper (Cu), Iron (Fe), Magnesium (Mg), Silicon (Si), Zinc (Zn). Please note that multi-element lamps are not designed to be used in a furnace system. In addition the analysis of Aluminum and Silicon require the use of a nitrous oxide-acetylene flame. Every genuine PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume — and the longer the lamp lifetime. Note: Lumina hollow cathode lamps may also be used with all earlier AA spectrometers that