Learn More
Sorbent Technologies Inc Alumina for TLC, 5-25um, w/Gypsum and UV254, 1kg
SDP
Catalog No. 501138788
Encompass_Preferred
Alumina for TLC with Gypsum and fluorescent indicator (UV254) has particle distribution of 5-25um, and a quantity of 1kg. This alumina is used for the making of analytical TLC plates.
Catalog No. 50-113-8788 Supplier Sorbent Technologies Inc Supplier No. 163501
Alumina for TLC with Gypsum and fluorescent indicator (UV254) has particle distribution of 5-25um, and a quantity of 1kg. This alumina is used for the making of analytical TLC plates.