Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1764662
Encompass_Preferred
Aluminum housed mantle for 1 gallon paint can to 400 degrees C maximum internal operating temperature, 590W, 115V
Catalog No. NC1764662 Supplier Glas Col LLC Supplier No. 100B CH097
 Restricted Item
Add to Cart
Add to Cart
Aluminum housed mantle for 1 gallon paint can to 400 degrees C maximum internal operating temperature, 590W, 115V