Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1764661
Encompass_Preferred
Aluminum housed mantle for 1 quart paint can, to 400 degrees C maximum internal operating temperature, 325W, 115V
Catalog No. NC1764661 Supplier Glas Col LLC Supplier No. 100B CH095
Aluminum housed mantle for 1 quart paint can, to 400 degrees C maximum internal operating temperature, 325W, 115V