Learn More
Sorbent Technologies Inc Alumina N TLC Plates, aluminum backed, 200um, 20x20cm, 25/pk
SDP
Catalog No. 501138485
Encompass_Preferred
Alumina Neutral (pH 6-8) analytical, aluminum backed TLC plates. The coating thickness is 200um and dimensions are 20x20cm. There are 25 plates per pack. Applications include: terpenes, alkaloids, steroids, aliphatic and aromatic compounds. To ensure sufficient adherence an inert organic binder is added to the aqueous suspension for coating. The aluminum oxide layer has a specific surface of about 200 m2/g (BET) and a pore size of about 60A. To activate the aluminum oxide layer, we recommend heating the TLC plates for 10 minutes at 120C.
Catalog No. 50-113-8485 Supplier Sorbent Technologies Inc Supplier No. 0234026
Alumina Neutral (pH 6-8) analytical, aluminum backed TLC plates. The coating thickness is 200um and dimensions are 20x20cm. There are 25 plates per pack. Applications include: terpenes, alkaloids, steroids, aliphatic and aromatic compounds. To ensure sufficient adherence an inert organic binder is added to the aqueous suspension for coating. The aluminum oxide layer has a specific surface of about 200 m2/g (BET) and a pore size of about 60A. To activate the aluminum oxide layer, we recommend heating the TLC plates for 10 minutes at 120C.