Learn More
Electron Microscopy Sciences Aluminum Oxide Abrasive Discs 8", PSA backing
SDP
Supplier:  Electron Microscopy Sciences 503589
Encompass_Preferred
Alumina is widely used in sample grinding and polishing. Our Aluminum Oxide Abrasive film with a PSA back or a plain back are ready to use in all sample polishing applications.
Backing: PSA
Catalog No. 50-997-296