Learn More
Cole-Parmer Cole-Parmer Aluminum Smooth-Walled Weighing Dishes, 150 mL, 100/Pk
Catalog No. NC1788951 Shop All Cole-Parmer Products
Encompass_Preferred
Smooth-sided, flat bottom, multipurpose "utility" dish. Oil-free and non-coated, no tab; ideal for storage, dispensing, general purpose sampling and testing. Dish contour facilitates stacking and dispensing. May be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical.
Catalog No. NC1788951 Supplier Cole-Parmer Supplier No. 0101832
Smooth-sided, flat bottom, multipurpose "utility" dish. Oil-free and non-coated, no tab; ideal for storage, dispensing, general purpose sampling and testing. Dish contour facilitates stacking and dispensing. May be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical.