Learn More
Selleck Chemical LLC AL082D06
SDP
Catalog No. 5000002913
Encompass_Preferred
AL082D06 is a nonsteroidal glucocorticoid receptor (GR) antagonist with no detectable binding affinity for the highly related receptors for mineralocorticoids androgens estrogens and progestins
Catalog No. 50-000-02913 Supplier Selleck Chemical LLC Supplier No. S66082MG
AL082D06 is a nonsteroidal glucocorticoid receptor (GR) antagonist with no detectable binding affinity for the highly related receptors for mineralocorticoids androgens estrogens and progestins