Learn More
Hach Company AL110 Salt-Bridge Dbl Junction
Catalog No. 5000207223 Shop All Hach Company Products
Encompass
Radiometer Analytical AL110 Replacement Salt-Bridge Double Liquid Junction (length 250 mm-ceramic junction)The Radiometer AL110 salt-bridge junction is a double-junction replacement salt bridge for 7 5 or 8 mm diameter reference electrodes The AL110 has a total length of 250 mm During measurement the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction
Catalog No. 50-002-07223 Supplier Hach Company Supplier No. B40A550
Radiometer Analytical AL110 Replacement Salt-Bridge Double Liquid Junction (length 250 mm-ceramic junction)The Radiometer AL110 salt-bridge junction is a double-junction replacement salt bridge for 7 5 or 8 mm diameter reference electrodes The AL110 has a total length of 250 mm During measurement the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction