Learn More
Hach Company AL120 Salt-Bridge Dbl Junction
Catalog No. 5000207165 Shop All Hach Company Products
Encompass
Radiometer Analytical AL120 Replacement Salt-Bridge Double Liquid Junction (x2 ground joints-length 138 mm-ceramic)The Radiometer AL120 salt-bridge junction is a double-junction replacement salt bridge for reference electrodes with a ground joint The AL120 has a total length of 138 mm During measurement the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction
Catalog No. 50-002-07165 Supplier Hach Company Supplier No. B40A610
Radiometer Analytical AL120 Replacement Salt-Bridge Double Liquid Junction (x2 ground joints-length 138 mm-ceramic)The Radiometer AL120 salt-bridge junction is a double-junction replacement salt bridge for reference electrodes with a ground joint The AL120 has a total length of 138 mm During measurement the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction