Learn More
Medchemexpress LLC β-Ala-Gly-Arg-pNA | 858971-42-3 | 99.39% | 422.44 | 5 MG
SDP
Catalog No. 5000422494
Encompass_Preferred
β-Ala-Gly-Arg-pNA is a chromogenic substrate of thrombin. It exhibits a strong absorbance at 405 nm due to its pNA group.
  • Functions as a chromogenic substrate.
  • Used as a thrombin substrate.
  • Features strong absorbance at 405 nm.
Catalog No. 50-004-22494 Supplier Medchemexpress LLC Supplier No. HYP100945MG
β-Ala-Gly-Arg-pNA is a chromogenic substrate of thrombin. It exhibits a strong absorbance at 405 nm due to its pNA group.
  • Functions as a chromogenic substrate.
  • Used as a thrombin substrate.
  • Features strong absorbance at 405 nm.