Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-Ala-Gly-Arg-pNA | 858971-42-3 | 99.4% | 422.44 | 10 MG
SDP

Catalog No. 5000423419
Encompass_Preferred

β-Ala-Gly-Arg-pNA is a chromogenic substrate of thrombin, exhibiting strong absorbance at 405 nm. It is intended for research use only and appears as an off-white to light yellow solid.

  • Chromogenic substrate of thrombin
  • Exhibits strong absorbance at 405 nm
  • For research use only
  • Appearance: off-white to light yellow solid
  • Stable for 2 years at -80°C and 1 year at -20°C as powder
  • Stable for 6 months at -80°C and 1 month at -20°C in solvent

Catalog No. 50-004-23419 Supplier Medchemexpress LLC Supplier No. HYP1009410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Ala-Gly-Arg-pNA is a chromogenic substrate of thrombin, exhibiting strong absorbance at 405 nm. It is intended for research use only and appears as an off-white to light yellow solid.

  • Chromogenic substrate of thrombin
  • Exhibits strong absorbance at 405 nm
  • For research use only
  • Appearance: off-white to light yellow solid
  • Stable for 2 years at -80°C and 1 year at -20°C as powder
  • Stable for 6 months at -80°C and 1 month at -20°C in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.