Learn More
CPC Scientific H-Ala-Leu-Ala-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific TAT004A
Encompass
A peptidase labile sequence that links drugs to proteins or lipophilic carriers. ONE-LETTER SEQUENCE: ALALMOLECULAR FORMULA: C18H34N4O5MOLECULAR WEIGHT:386.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [84676-48-2], RESEARCH AREA: AntimicrobialREFERENCES: A. Trouet, et al., Targeting of Drugs, 19-30 New York (1982); M. Studer, et al., Bioconjug. Chem. 3, 424-429 (1992); A.J. Versluis, et al., Pharmaceut. Res. 15, 531-537 (1998)
Catalog No. 50-211-0543