Learn More
Carbosynth LLC. β-Ala-Lys(AMCA) (5 mg vial), CAS# 180342-52-3, MF= C21H28N4O6
SDP
Catalog No. 501952415
Encompass_Preferred
β-Alanyl-Nε-(7-Amino-4-Methyl-2-Oxo-2H-1-Benzopyran-3-Acetyl)-L-Lysine, Application: Fluorescent Tracer for Oligopeptide Transporters, MW: 432.47
Catalog No. 50-195-2415 Supplier Carbosynth LLC. Supplier No. PBP3412V5MG
β-Alanyl-Nε-(7-Amino-4-Methyl-2-Oxo-2H-1-Benzopyran-3-Acetyl)-L-Lysine, Application: Fluorescent Tracer for Oligopeptide Transporters, MW: 432.47