Learn More
Medchemexpress LLC Ala-parafluoroPhe-Arg-Cha-Cit-Tyr-NH2 TFA | 99.8% | 883.02 (free base) | 5 MG
SDP
Catalog No. 5000398904
Encompass_Preferred
Ala-parafluoroPhe-Arg-Cha-Cit-Tyr-NH2 TFA is a bioactive peptide. It acts as a selective Protease activating receptor 1 (PAR-1) agonist over PAR-2. PAR-1 is a subfamily of G-protein coupled receptors that mediates the cellular effects of thrombin. Additionally, PAR-1, in coordination with PAR-4, regulates thrombin-induced hepatocellular carcinoma within the tumor environment.
  • Bioactive peptide
  • Selective Protease activating receptor 1 (PAR-1) agonist over PAR-2
  • Regulates thrombin-induced hepatocellular carcinoma within the tumor environment
  • For research use only
Catalog No. 50-003-98904 Supplier Medchemexpress LLC Supplier No. HYP5372A5MG
Ala-parafluoroPhe-Arg-Cha-Cit-Tyr-NH2 TFA is a bioactive peptide. It acts as a selective Protease activating receptor 1 (PAR-1) agonist over PAR-2. PAR-1 is a subfamily of G-protein coupled receptors that mediates the cellular effects of thrombin. Additionally, PAR-1, in coordination with PAR-4, regulates thrombin-induced hepatocellular carcinoma within the tumor environment.
  • Bioactive peptide
  • Selective Protease activating receptor 1 (PAR-1) agonist over PAR-2
  • Regulates thrombin-induced hepatocellular carcinoma within the tumor environment
  • For research use only