Learn More
Medchemexpress LLC [Ala11,D-Leu15]-Orexin B(human) | 532932-99-3 | 98.3% | 2857.26 | 5 MG
SDP
Catalog No. 5000424569
Encompass_Preferred
Ala11, D-Leu15-Orexin B(human) is a potent and selective orexin-2 receptor (OX2) agonist. This peptide demonstrates a 400-fold selectivity for OX2 (EC50=0.13 nM) over OX1 (52 nM), highlighting its specific action on the orexin system. It is provided with a purity of 98.3% and is intended for research use only.
  • Potent and selective OX2 agonist.
  • High selectivity for OX2 over OX1.
  • For research use only.
  • High purity.
Catalog No. 50-004-24569 Supplier Medchemexpress LLC Supplier No. HYP13405MG
Ala11, D-Leu15-Orexin B(human) is a potent and selective orexin-2 receptor (OX2) agonist. This peptide demonstrates a 400-fold selectivity for OX2 (EC50=0.13 nM) over OX1 (52 nM), highlighting its specific action on the orexin system. It is provided with a purity of 98.3% and is intended for research use only.
  • Potent and selective OX2 agonist.
  • High selectivity for OX2 over OX1.
  • For research use only.
  • High purity.