Learn More
Cayman Chemical Ala17-MCHtrIfluoroacetate 10mg
SDP
Catalog No. 502913458
Encompass_Preferred
An MCH-derived peptide and MCH receptor agonist; binds to MCH1 and MCH2 receptors in CHO cells expressing the human receptors (IC50s = 0.15 and 3.5 nM, respectively); induces calcium mobilization in HEK293 cells expressing the human receptors (EC50s = 17 and 54 nM, respectively)
Catalog No. 50-291-3458 Supplier Cayman Chemical Supplier No. 3526210 MG
An MCH-derived peptide and MCH receptor agonist; binds to MCH1 and MCH2 receptors in CHO cells expressing the human receptors (IC50s = 0.15 and 3.5 nM, respectively); induces calcium mobilization in HEK293 cells expressing the human receptors (EC50s = 17 and 54 nM, respectively)