Learn More
CPC Scientific H-Lys-Lys-Ala-Leu-Arg-Arg-Gln-Glu-Ala-Val-Asp-Ala-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP001A
Encompass
Highly specific and potent inhibitor of calmodulin-dependent protein Kinase II (IC50= 40 nM) that does not affect cycle AMP-dependent protein kinase, protein kinase C and calmodulin-dependent protein kinase IV.ONE-LETTER SEQUENCE: KKALRRQEAVDALMOLECULAR FORMULA: C64H116N22O19MOLECULAR WEIGHT:1497.74STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [167114-91-2], RESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0155