Learn More
Enzo Life Sciences (Ala97)-RII (81-99) (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP1180500
Encompass_Preferred
This sequence,DLDVPIPGRFDRRVSVAAE, is from cAMP-dependent protein kinase regulatory subunit type II (PKA). The phosphorylated peptide (BML-P160) can be used as a substrate for calcineurin (Km=25 µM, Vmax=6 µmol/min/mg). Alternative name: [Ala97] cAMP dependent protein kinase regulatory subunit type 2 (81 99). Purity: ≥97%. Long Term Storage: -20°C.
Catalog No. 50-201-1235