Learn More
Aviva Systems Biology Corp ALAD Antibody - N-terminal region (ARP41845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALAD. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ALAD. Alternate names: ALADH, MGC5057, PBGSApplicationWB. Species reactivity: Human
Catalog No. 50-284-959