Learn More
Aviva Systems Biology Corp ALAD Antibody - N-terminal region (ARP41656_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41656_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALAD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ALAD. Alternate names: ALADH, MGC5057, PBGSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57934