Learn More
Apexbio Technology LLC Alagebrium chloride 341028-37-3 200mg
SDP
Catalog No. 5000096254
Encompass_Preferred
Alagebrium chloride (CAS 341028-37-3) is a small molecule inhibitor that targets advanced glycation end-products (AGEs) by cleaving pre-existing AGE-derived protein cross-links AGEs are formed through non-enzymatic reactions between reducing sugars and proteins or lipids progressively accumulating with age and contributing to tissue stiffening and functional decline By disrupting AGE-mediated cross-linking alagebrium chloride has been studied for its potential to restore tissue elasticity and improve organ function This compound is utilized in research investigating age-related pathological processes and therapeutic strategies for diseases associated with elevated AGE accumulation
Catalog No. 50-000-96254 Supplier Apexbio Technology LLC Supplier No. A9915200
Alagebrium chloride (CAS 341028-37-3) is a small molecule inhibitor that targets advanced glycation end-products (AGEs) by cleaving pre-existing AGE-derived protein cross-links AGEs are formed through non-enzymatic reactions between reducing sugars and proteins or lipids progressively accumulating with age and contributing to tissue stiffening and functional decline By disrupting AGE-mediated cross-linking alagebrium chloride has been studied for its potential to restore tissue elasticity and improve organ function This compound is utilized in research investigating age-related pathological processes and therapeutic strategies for diseases associated with elevated AGE accumulation