Learn More
Pyramex™ Alair™ Safety Eyewear
Catalog No. 19155553 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Silver, Mirror
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-553 Silver, Mirror
19-155-550 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-155-553 Supplier Pyramex™ Supplier No. S3270S
Add to Cart
Add to Cart
This item is not returnable. View return policy
A stylish look that is 100% polycarbonate. These economical glasses have a hard coated lens-unlike most at this price
  • Economically priced to fit any budget
  • Incredibly light, resilient & durable protective eyewear for all day wear
  • Closefitting unitary front with dual 9.5 base curvature offers wide and unobstructed vision
  • Fashionable design resembles sunglasses, suitable for both men and women
  • Scratch-resistant, hard-coated polycarbonate lens filter out more than 99.9% of harmful UV rays
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Silver, Mirror
    Frame Color Silver
    Lens Material Polycarbonate