Learn More
Thermo Scientific™ alamarBlue™ Dye
Catalog No. 10230103 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-230-103 10 mL
10-230-102 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-230-103 Supplier Thermo Scientific™ Supplier No. Y00010
Add to Cart
Add to Cart
This item is not returnable. View return policy
Allows three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
Thermo Scientific™ alamarBlue provides unmatched sensitivity and versatility, allowing three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
alamarBlue, a resazurin-based product with extended incubation time, is stable for up to 5 days.
TRUSTED_SUSTAINABILITY
Specifications
Description alamarBlue indicator dye
Quantity 10 mL
How do alamarBlue reagent and PrestoBlue reagent differ from resazurin and C12-resazurin?
alamarBlue reagent and PrestoBlue reagent contain resazurin in a proprietary stabilizing formulation that allows for a convenient “mix, incubate, and read” protocol. PrestoBlue reagent is an improvement in the formulation of alamarBlue reagent that allows for much faster staining (typically 10 minutes vs. 1-4 hours to obtain a similar signal and sensitivity). C12-resazurin is a derivative of resazurin that has better cellular retention and thus allows for analysis on a flow cytometer and multiplexing with viability indicators and other biomarkers.